Learn More
R&D Systems™ Recombinant Mouse IL-33 Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT. |
| Gene ID (Entrez) | 77125 |
| Molecular Weight (g/mol) | 18 kDa |
| Name | IL-33 |
| Quantity | 10 μg |
| Source | E. coli-derived mouse IL-33 protein Ser109-Ile266 |
| Storage Requirements | 12 months from date of receipt, ≤ -20° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.