Learn More
R&D Systems™ Mouse/Rat GDF-15 Quantikine ELISA Kit

Description
Includes
Cell Culture Supernates (50μL), Serum (50μL), EDTA Plasma (50μL), Heparin Plasma (50μL), Urine (10μL)
Specifications
Specifications
| Assay Range | 7.80 - 500pg/mL |
| Assay Sensitivity | 2.2pg/mL |
| Conjugate | HRP |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine |
| Specificity | Natural Mouse and Rat GDF-15, Recombinant Mouse mature GDF-15, precursor GDF-15, and GDF-15/BMP-2 heterodimer. It does not detect monomeric Recombinant Mouse GDF-15. |
| Target Species | Mouse, Rat |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Sample Volume | Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Urine (10uL) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.