Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse/Rat PDGF-BB DuoSet ELISA

Catalog No. dy846405
Change view
Click to view available options
Quantity:
5 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY846405 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY846405 Supplier R&D Systems™ Supplier No. DY846405
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

For the development of sandwich ELISAs to measure natural and recombinant mouse and rat Platelet-derived Growth Factor BB (PDGF-BB)

The Mouse/Rat PDGF-BB DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse, rat PDGF-BB.

Specifications

Assay Range 62.50 to 4000 pg/mL
Detectable Analytes PDGF-BB
Conjugate Biotin
Immunoassay Kit Format 96-well Strip Plate
Product Type ELISA Kit
Sufficient For To run ELISAs on at least 5 x 96-well plates
Target Species Mouse, Rat
For Use With (Application) ELISA
Gene ID (Entrez) 5155
Host Species Mouse, Rat
Quantity 5 Plates
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.