Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse SIRP alpha/CD172a Fc Chimera Protein
SDP

Catalog No. 7154SA050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7154SA050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 7154SA050 Supplier R&D Systems™ Supplier No. 7154SA050

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by the ability of the immobilized protein to support the adhesion of mouse red blood cells.

The ED50 for this effect is 0.4-1.6 μg/mL.

Optimal dilutions should be determined by each laboratory for each application.

Specifications

Accession Number P97797
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 140885
Molecular Weight (g/mol) M.W. (Observed): 115-125 kDa, reducing conditions; M.W. (theoretical): 65.1 kDa (monomer)
Name SIRP alpha/CD172a Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse SIRP alpha/CD172a protein Mouse SIRP alpha/CD172a (Met1-Asn373) (Gly365Asp) (N-terminus) Accession # P97797 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol Sirpa
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein SIRP alpha/CD172a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.