Learn More
R&D Systems™ Proteome Profiler Human Chemokine Array Kit
GREENER_CHOICESDP
Catalog No. ARY017
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
ARY017 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. ARY017 Supplier R&D Systems™ Supplier No. ARY017
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
The Proteome Profiler Human Chemokine Array Kit requires no specialized equipment and eliminates the need for multiple Western blot experiments.
The Proteome Profiler Human Chemokine Array Kit contains 4 membranes - each spotted in duplicate with 31 different chemokine antibodies.
Specifications
Quantity 1 Kit
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.