Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat IFN-gamma Protein

Catalog No. p-4722624
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Tris, NaCl and PEG with BSA as a carrier protein.
Supplied as a 0.2μm filtered solution in Tris, NaCl and PEG 3350.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
585IF100CF Supplied as a 0.2μm filtered solution in Tris, NaCl and PEG 3350.
585IF100 Lyophilized from a 0.2μm filtered solution in Tris, NaCl and PEG with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 585IF100CF Supplier R&D Systems™ Supplier No. 585IF100/CF
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Rat IFN-gamma Protein is derived from E. coli. The Recombinant Rat IFN-gamma Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Supplied as a 0.2μm filtered solution in Tris, NaCl and PEG 3350.
Gene ID (Entrez) 25712
Molecular Weight (g/mol) 15.5 kDa
Name IFN-gamma
Quantity 100 μg
Source E. coli-derived rat IFN-gamma protein Glu23-Cys156,with an N-terminal Met
Storage Requirements 3 months from date of receipt, 2 to 8° C as supplied.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.