Learn More
R&D Systems™ Resazurin
GSA_VA
Catalog No. ar002
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
AR002 100 mL
1 options
This item is not returnable. View return policy
Catalog No. AR002 Supplier R&D Systems™ Supplier No. AR002
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Simple and quantitative method for measuring cell proliferation, viability and cytotoxicity
  • Nontoxic, water soluble, redox-sensitive dye that changes from its blue/non-fluorescent state to pink/highly-fluorescent upon reduction to resorufin by viable cells
  • Fluorescence can be read using 544nm excitation and 590nm emission wavelengths, or absorbance can be read using a spectrophotometer set at 570nm
  • Resazurin may be added at any time during the culture period
  • For measurement of cell proliferation, it is best to add Resazurin during log phase growth
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 100 mL
Content And Storage Store the unopened product at 2 - 8° C. Protect from light. Do not use past expiration date.