Learn More
R&D Systems™ Streptavidin-Allophycocyanin
Description
- Secondary reagent in immunofluorescent assays using biotinylated primary labeling reagents
- 1mL of SA-APC at a concentration of 10μg/mL
- Allophycocyanin has an absorption spectrum from 620 to 650nm and has optimal emission at 660 to 670nm
Applications:
Flow Cytometry
Specifications
Specifications
| Formulation | Ready-to-use Liquid |
| Quantity | 100 Tests |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.