Learn More
R&D Systems™ Tyrosine Phosphatase Substrate I, DADEY(PO3)LIPQQG

Description
- Enzymatic cleavage of this substrate liberates inorganic phosphate, which can be measured in dye binding assays, such as the malachite green method
- Synthetic phosphopeptide corresponding to amino acids 988 - 998 of the mature EGF receptor
- Derived from an autophosphorylation site at Y992 of EGFR
- Purity 98% based on HPLC analysis
- Dissolves in water
Specifications
Specifications
| Form | Lyophilized |
| Quantity | 1 mg |
| Format | Lyophilized from a stock solution in distilled water. |
| Molecular Weight (g/mol) | Predicted MW: 1328.32 Da |
| Product Type | Purity: >95%, by HPLC, Source: Synthetic |
| Storage Requirements | Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquotted and stored -20° C to -70° C in a manual defrost freezer for one month. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.