Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Z-FR-AMC Fluorogenic Peptide Substrate
SDP

Catalog No. ES009
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES009 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. ES009 Supplier R&D Systems™ Supplier No. ES009

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Substrate for Papain

  • Hydrolysis of Arg-AMC amide bond releases 7-amino-4-methyl coumarin (AMC), a highly fluorescent group
  • A fluorescence plate reader with excitation at 380nm and emission at 460nm is recommended for the measurement of the enzymatic activity
  • Often used in characterization of cysteine protease inhibitors
  • Purity >99% by thin-layer chromatography

Specifications

Form Liquid
For Use With (Application) Bioactivity
Quantity 50 mg
Format Supplied as a stock solution in dimethyl sulfoxide (DMSO) with a concentration of 100 mg/mL by mass or 157.4 mM with consideration of the peptide content.
Molecular Weight (g/mol) Theoretical Molecular weight: 612.6 Da
Product Type Z-Phe-Arg-AMC
Storage Requirements No Stability Data Storage is set to N/A for ProdCode
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.