Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-D2PM | 22348-32-9 | 253.34 | 25 G
SDP

Catalog No. 5000449862
Encompass_Preferred

(R)-D2PM, also known as (R)-Diphenylprolinol, is a pyrrolidine analog supplied as a white to off-white solid with 99.9% purity. It is for research use only and requires specific handling and storage to maintain stability.

  • High purity: 99.9%
  • Appearance: White to off-white solid
  • Storage: 4°C, protect from light, stored under nitrogen
  • In solvent storage: -80°C for 6 months; -20°C for 1 month, protected from light, stored under nitrogen
  • Melting point: 77-80°C
  • Boiling point: 411.3°C at 760 mmHg
  • Solubility: 10 mg/mL in DMSO
  • Causes skin and serious eye irritation
  • May cause respiratory irritation

Catalog No. 50-004-49862 Supplier Medchemexpress LLC Supplier No. 25GHYW007880
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-D2PM, also known as (R)-Diphenylprolinol, is a pyrrolidine analog supplied as a white to off-white solid with 99.9% purity. It is for research use only and requires specific handling and storage to maintain stability.

  • High purity: 99.9%
  • Appearance: White to off-white solid
  • Storage: 4°C, protect from light, stored under nitrogen
  • In solvent storage: -80°C for 6 months; -20°C for 1 month, protected from light, stored under nitrogen
  • Melting point: 77-80°C
  • Boiling point: 411.3°C at 760 mmHg
  • Solubility: 10 mg/mL in DMSO
  • Causes skin and serious eye irritation
  • May cause respiratory irritation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.