Learn More
Cayman Chemical R-(−)-Deprenyl (hydrochloride), Purity: ≥98%, CAS: 14611-52-0, 100 mg
SDP
Catalog No. 502969159
Encompass_Preferred
A selective, reversible inhibitor of MAO-B (Ki = 0.091 μM) over MAO-A (Ki = 9.06 μM); may ameliorate depression, Parkinson’s disease, Alzheimer’s disease, Huntington’s disease, and stroke
Catalog No. 50-296-9159 Supplier Cayman Chemical Supplier No. 15046100 MG
A selective, reversible inhibitor of MAO-B (Ki = 0.091 μM) over MAO-A (Ki = 9.06 μM); may ameliorate depression, Parkinson’s disease, Alzheimer’s disease, Huntington’s disease, and stroke