Learn More
Medchemexpress LLC (R)-donepezil | 142698-19-9 | MFCD21396234 | 99.9% | 379.49 | C24H29NO3 | 5 MG
SDP
Catalog No. 5000222779
Encompass_Preferred
(R)-Donepezil is the R-enantiomer of donepezil, a specific and potent acetylcholinesterase (AChE) inhibitor used in biochemical and pharmacological research. Supplied as a solid with high chemical and enantiomeric purity, it is intended for stereochemistry-focused assays and mechanistic studies. Analytical documentation and spectral data are available to support characterization.
Catalog No. 50-002-22779 Supplier Medchemexpress LLC Supplier No. HY1368385MG
(R)-Donepezil is the R-enantiomer of donepezil, a specific and potent acetylcholinesterase (AChE) inhibitor used in biochemical and pharmacological research. Supplied as a solid with high chemical and enantiomeric purity, it is intended for stereochemistry-focused assays and mechanistic studies. Analytical documentation and spectral data are available to support characterization.