Learn More
Medchemexpress LLC (R)-DRF053 dihydrochloride | 1241675-76-2 | 99.3% | 490.43 g/mol | C23H29Cl2N7O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1033835MG
Encompass_Preferred
(R)-DRF053 dihydrochloride is a small-molecule kinase inhibitor (dihydrochloride salt) that potently inhibits casein kinase 1 (CK1) and cyclin-dependent kinases CDK1 and CDK5. It has been used in cellular studies to reduce CK1-dependent amyloid-beta production and is provided in small quantities for research use.
  • Potent CK1 inhibition (IC50 14 nM) with additional CDK1 and CDK5 activity (IC50 220 nM and 80 nM).
  • Supplied as the dihydrochloride salt for improved solubility in aqueous media.
  • High purity suitable for research use (99.3%).
  • Available in small research quantities (1 mg, 5 mg, 10 mg).
  • Recommended storage sealed at 4°C; in solvent, store at -80°C (up to 6 months) or -20°C (up to 1 month).
Catalog No. 50-002-44977