Learn More
Medchemexpress LLC (R,E)-N-(Cyclopropylmethyl)-N-methyl-3,6-diphenylhex-5-en-3-amine hydrochloride | 99.0% | 355.94 | 1 ML
SDP
Catalog No. 5000346486
Encompass_Preferred
(+)-Igmesine hydrochloride is a white to off-white solid that has been tested and complies with given specifications. It is suitable for various laboratory applications requiring precise chemical properties.
  • Stored at -20°C, protected from light, under nitrogen.
  • In solvent, stored at -80°C for 6 months or -20°C for 1 month, protected from light, under nitrogen.
  • High purity of 99.0% (HPLC).
Catalog No. 50-003-46486 Supplier Medchemexpress LLC Supplier No. HY10851510MG
(+)-Igmesine hydrochloride is a white to off-white solid that has been tested and complies with given specifications. It is suitable for various laboratory applications requiring precise chemical properties.
  • Stored at -20°C, protected from light, under nitrogen.
  • In solvent, stored at -80°C for 6 months or -20°C for 1 month, protected from light, under nitrogen.
  • High purity of 99.0% (HPLC).