Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-eIF4A3-IN-2 | 2095484-82-3 | 95.0% | 602.70 | C25H19Br2ClN4O2 | 10 MG
SDP

Catalog No. 5000298495
Encompass_Preferred

(R)-eIF4A3-IN-2 is the R-enantiomer of eIF4A3-IN-2 and a less-active stereoisomer of a selective, noncompetitive inhibitor of eukaryotic initiation factor 4A-3 (eIF4A3). The compound is supplied as a solid and is provided with recommended storage and solubility information for laboratory handling.

  • R enantiomer with reduced activity compared to the parent compound.
  • Selective, noncompetitive inhibitor of eIF4A3; parent IC50 = 110 nM.
  • CAS number 2095484-82-3.
  • Molecular formula C25H19Br2ClN4O2 and molecular weight 602.70.
  • Purity approximately 95.0% as supplied.
  • Physical form: solid, white to light yellow.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solution -80°C (2 years) or -20°C (1 year).
  • Solubility: soluble in DMSO (~50 mg/mL; ultrasonic assistance recommended).

Catalog No. 50-002-98495 Supplier Medchemexpress LLC Supplier No. HY4391310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-eIF4A3-IN-2 is the R-enantiomer of eIF4A3-IN-2 and a less-active stereoisomer of a selective, noncompetitive inhibitor of eukaryotic initiation factor 4A-3 (eIF4A3). The compound is supplied as a solid and is provided with recommended storage and solubility information for laboratory handling.

  • R enantiomer with reduced activity compared to the parent compound.
  • Selective, noncompetitive inhibitor of eIF4A3; parent IC50 = 110 nM.
  • CAS number 2095484-82-3.
  • Molecular formula C25H19Br2ClN4O2 and molecular weight 602.70.
  • Purity approximately 95.0% as supplied.
  • Physical form: solid, white to light yellow.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solution -80°C (2 years) or -20°C (1 year).
  • Solubility: soluble in DMSO (~50 mg/mL; ultrasonic assistance recommended).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.