Learn More
Medchemexpress LLC HY-108414 10mg Medchemexpress, (R)-Equol CAS:221054-79-1 Purity:>98%
SDP
Catalog No. 501968542
Encompass_Preferred
Medchemexpress, HY-108414 10mg (R)-Equol CAS:221054-79-1 (R)-Equol is an agonist of both ERα and ERβ with Kis of 27.4 and 15.4 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8542 Supplier Medchemexpress LLC Supplier No. HY10841410MG
Medchemexpress, HY-108414 10mg (R)-Equol CAS:221054-79-1 (R)-Equol is an agonist of both ERα and ERβ with Kis of 27.4 and 15.4 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.