Learn More
Medchemexpress LLC (R)-Ethyl 2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate | 1146954-88-2 | MFCD18071557 | 97.0% | 233.26 | C10H19NO5 | 10 G
SDP
Catalog No. 5000299655
Encompass_Preferred
(R)-Ethyl 2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate is the R-enantiomer of a Boc-protected serine ethyl ester used as a protected amino acid building block in synthetic and medicinal chemistry. Supplied as a solid for research use, it enables incorporation of protected serine residues in peptide synthesis and small-molecule routes.
  • R-enantiomer for stereospecific synthesis.
  • Boc-protected amino group for orthogonal protection.
  • Ethyl ester provides protected carboxyl functionality.
  • High purity (97.0%) suitable for research applications.
  • Solid, white to off-white form for easy handling.
  • Storage stability: powder stable at -20°C and 4°C for extended periods.
  • Documentation available: datasheet, COA, and SDS.
Catalog No. 50-002-99655 Supplier Medchemexpress LLC Supplier No. HYW02253610G
Add to Cart
Add to Cart
(R)-Ethyl 2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate is the R-enantiomer of a Boc-protected serine ethyl ester used as a protected amino acid building block in synthetic and medicinal chemistry. Supplied as a solid for research use, it enables incorporation of protected serine residues in peptide synthesis and small-molecule routes.
  • R-enantiomer for stereospecific synthesis.
  • Boc-protected amino group for orthogonal protection.
  • Ethyl ester provides protected carboxyl functionality.
  • High purity (97.0%) suitable for research applications.
  • Solid, white to off-white form for easy handling.
  • Storage stability: powder stable at -20°C and 4°C for extended periods.
  • Documentation available: datasheet, COA, and SDS.