Learn More
Medchemexpress LLC (R)-ethyl 2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate | 1146954-88-2 | MFCD18071557 | 97.0% | 233.26 g/mol | C10H19NO5 | 100 G
SDP
Catalog No. 5000301587
Encompass_Preferred
(R)-Ethyl 2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate is the R-enantiomer of an N-Boc protected serine ethyl ester used as a synthetic intermediate in organic and peptide chemistry. It serves as a stereochemically defined building block for incorporation of protected serine residues and for selective functionalization in multi-step syntheses.
  • R-configured N-Boc protected serine ethyl ester.
  • Useful intermediate for peptide synthesis and amino acid derivatization.
  • High purity suitable for research applications.
  • Protected hydroxyl and amino groups allow selective transformations.
  • Compatible with common coupling and deprotection protocols.
Catalog No. 50-003-01587 Supplier Medchemexpress LLC Supplier No. HYW022536100G
(R)-Ethyl 2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate is the R-enantiomer of an N-Boc protected serine ethyl ester used as a synthetic intermediate in organic and peptide chemistry. It serves as a stereochemically defined building block for incorporation of protected serine residues and for selective functionalization in multi-step syntheses.
  • R-configured N-Boc protected serine ethyl ester.
  • Useful intermediate for peptide synthesis and amino acid derivatization.
  • High purity suitable for research applications.
  • Protected hydroxyl and amino groups allow selective transformations.
  • Compatible with common coupling and deprotection protocols.