Learn More
Medchemexpress LLC (R)-Ethyl 2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate | 1146954-88-2 | MFCD18071557 | 97.0% | 233.26 g/mol | C10H19NO5 | 25 G
SDP
Catalog No. 5000303016
Encompass_Preferred
(R)-Ethyl 2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate is the R-enantiomer of a Boc-protected serine ethyl ester, intended as a building block for peptide synthesis and small-molecule assembly. It is supplied as a powder with specified purity and storage recommendations for research use.
  • Enantiomerically pure R-configuration.
  • Boc-protected amino acid derivative for peptide and organic synthesis.
  • High reported purity: 97.0%.
  • Offered in multiple sizes for lab-scale flexibility.
  • Recommended storage preserves stability and shelf life.
Catalog No. 50-003-03016 Supplier Medchemexpress LLC Supplier No. 25GHYW022536
(R)-Ethyl 2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate is the R-enantiomer of a Boc-protected serine ethyl ester, intended as a building block for peptide synthesis and small-molecule assembly. It is supplied as a powder with specified purity and storage recommendations for research use.
  • Enantiomerically pure R-configuration.
  • Boc-protected amino acid derivative for peptide and organic synthesis.
  • High reported purity: 97.0%.
  • Offered in multiple sizes for lab-scale flexibility.
  • Recommended storage preserves stability and shelf life.