Learn More
Apexbio Technology LLC (R)-(+)-Etomoxir sodium salt,10mg CAS# 828934-41-4
SDP
Catalog No. 502239978
Encompass_Preferred
(R)-(+)-Etomoxir sodium salt is an irreversible inhibitor of carnitine palmitoyltransferase 1(CPT-1)[1]. Etomoxir (100 μM) has no effect on T cells cultured in high glucose. In contrast, there is a significant increase in apoptosis in etomoxir-treated cultures stimulated with antigen under low glucose conditions[2]. C57BL/6 mice treated with Etomoxir (15 mg/kg, i.p) reduce the infiltration of immune cells in the central nervous system. Only a small number of macrophages, activated microglia or T cells are present, while reducing the inflammatory response and Demyelinating reaction[2]. References:[1]. Rupp H, Zarain-Herzberg A, Maisch B. The Use of Partial Fatty Acid Oxidation Inhibitors for Metabolic Therapy of Angina Pectoris and Heart Failure. Herz, 2002, 27(7): 621-636.Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-223-9978 Supplier Apexbio Technology LLC Supplier No. B767910
(R)-(+)-Etomoxir sodium salt is an irreversible inhibitor of carnitine palmitoyltransferase 1(CPT-1)[1]. Etomoxir (100 μM) has no effect on T cells cultured in high glucose. In contrast, there is a significant increase in apoptosis in etomoxir-treated cultures stimulated with antigen under low glucose conditions[2]. C57BL/6 mice treated with Etomoxir (15 mg/kg, i.p) reduce the infiltration of immune cells in the central nervous system. Only a small number of macrophages, activated microglia or T cells are present, while reducing the inflammatory response and Demyelinating reaction[2]. References:[1]. Rupp H, Zarain-Herzberg A, Maisch B. The Use of Partial Fatty Acid Oxidation Inhibitors for Metabolic Therapy of Angina Pectoris and Heart Failure. Herz, 2002, 27(7): 621-636.Other sizes are also available. Please inqury us for quote.