Learn More
TARGETMOL CHEMICALS INC R-EU-1180-453 25MG
SDP
Catalog No. 502878222
Encompass_Preferred
Also available in 50 mg, 100 mg and bulk. Please contact Fisher for quotes. R-(+)-EU-1180-453 is a novel GluN2C/D selective NMDAR-positive modulator for the study of neurological disorders. Purity 100%
Catalog No. 50-287-8222 Supplier TARGETMOL CHEMICALS INC Supplier No. T6961125MG
Also available in 50 mg, 100 mg and bulk. Please contact Fisher for quotes. R-(+)-EU-1180-453 is a novel GluN2C/D selective NMDAR-positive modulator for the study of neurological disorders. Purity 100%