Learn More
Medchemexpress LLC (R)-Exatecan Intermediate 1 | 110351-91-2 | 99.9% | 263.25 | 250 MG
SDP
Catalog No. 5000402161
Encompass_Preferred
(R)-Exatecan Intermediate 1 is a crucial intermediate used in the synthesis of Camptothecin derivatives, and it can be utilized to prepare Exatecan mesylate. This intermediate facilitates the synthesis of Exatecan, a compound known for its low sensitivity to multidrug resistance and its significant value in anti-tumor research.
  • Key intermediate in the synthesis of Camptothecin derivatives.
  • Can be used to prepare Exatecan mesylate.
  • Enables the synthesis of Exatecan.
  • Exatecan exhibits low sensitivity to multidrug resistance.
  • Valuable for anti-tumor research.
  • Available in solid and solution forms.
  • High purity: 99.95%.
Catalog No. 50-004-02161 Supplier Medchemexpress LLC Supplier No. 250MGHY42487B
(R)-Exatecan Intermediate 1 is a crucial intermediate used in the synthesis of Camptothecin derivatives, and it can be utilized to prepare Exatecan mesylate. This intermediate facilitates the synthesis of Exatecan, a compound known for its low sensitivity to multidrug resistance and its significant value in anti-tumor research.
  • Key intermediate in the synthesis of Camptothecin derivatives.
  • Can be used to prepare Exatecan mesylate.
  • Enables the synthesis of Exatecan.
  • Exatecan exhibits low sensitivity to multidrug resistance.
  • Valuable for anti-tumor research.
  • Available in solid and solution forms.
  • High purity: 99.95%.