Learn More
Medchemexpress LLC HY-15187A 5mg Medchemexpress, (R)-Filanesib CAS:885060-08-2 Purity:>98%
SDP
Catalog No. 502081688
Encompass_Preferred
Medchemexpress, HY-15187A 5mg (R)-Filanesib CAS:885060-08-2 (R)-Filanesib ((R)-ARRY-520) is the R-enantiomer of Filanesib (HY-15187). Filanesib is a synthetic kinesin spindle protein (KSP) inhibitor with an IC50 of 6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1688 Supplier Medchemexpress LLC Supplier No. HY15187A5MG
Medchemexpress, HY-15187A 5mg (R)-Filanesib CAS:885060-08-2 (R)-Filanesib ((R)-ARRY-520) is the R-enantiomer of Filanesib (HY-15187). Filanesib is a synthetic kinesin spindle protein (KSP) inhibitor with an IC50 of 6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.