Learn More
eMolecules​ 725728-43-8 | (R)-Fmoc-2-amino-2-methyl-3-tritylsulfanyl-propionic acid | Chem-Impex599.750 | C38H33NO4S | 98.000 | CC(CSC(c1ccccc1)(c1ccccc1)c1ccccc1)(NC(=O)OCC1c2ccccc2-c2ccccc12)C(O)=O | 250mg | 386904310
Catalog No. NC1721130 Shop All eMolecules Inc. Products
Encompass_Preferred
(R)-Fmoc-2-amino-2-methyl-3-tritylsulfanyl-propionic acid | Chem-Impex | 725728-43-8599.750 | C38H33NO4S | 98.000 | CC(CSC(c1ccccc1)(c1ccccc1)c1ccccc1)(NC(=O)OCC1c2ccccc2-c2ccccc12)C(O)=O | 250mg | 386904310
Catalog No. NC1721130 Supplier eMolecules​ Supplier No. 386904310
(R)-Fmoc-2-amino-2-methyl-3-tritylsulfanyl-propionic acid | Chem-Impex | 725728-43-8599.750 | C38H33NO4S | 98.000 | CC(CSC(c1ccccc1)(c1ccccc1)c1ccccc1)(NC(=O)OCC1c2ccccc2-c2ccccc12)C(O)=O | 250mg | 386904310