Learn More
Abcam (R)-FSL-1, TLR2/6 agonist, 100UG
Catalog No. 502705465
Encompass_Preferred
MW 1666.16 g/mol. Toll-like receptor 2/6 (TLR2/6) agonist. Synthetic lipoprotein. Represents N-terminal part of Mycoplasma salivarium LP44. NF-κB and MAPK pathway activator. Active in vivo. Rstereoisomer of FSL-1 (ab144863).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5465 Supplier Abcam Supplier No. AB144864100UG
MW 1666.16 g/mol. Toll-like receptor 2/6 (TLR2/6) agonist. Synthetic lipoprotein. Represents N-terminal part of Mycoplasma salivarium LP44. NF-κB and MAPK pathway activator. Active in vivo. Rstereoisomer of FSL-1 (ab144863).
The product is subject to the following: Abcam Restricted Use Statement