Learn More
AdipoGen R-FSL-1
SDP
Supplier:  AdipoGen AGCP30010C100
Encompass_Preferred
Chemical. Formula C84H140N14O18S. MW 1666.2. Synthetic. R-FSL-1 R-Pam2CGDPKHPKSF is a synthetic lipoprotein that represents the N-terminal part of the 44kDa lipoprotein LP44 of Mycoplasma salivarium. The naturally occurring R-stereoisomer is biologically more active than the S-stereoisomer. Stimulator of TLR2/TLR6. Inducer of TNF-alpha production in macrophages. Upregulates proinflammatory cytokines. Activator of the proinflammatory transcription factor NF-kappaB.
Catalog No. 50-256-2454