Learn More
Medchemexpress LLC HY-100742A 10mg , (R)-GNE-140 CAS:2003234-63-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100742A10MG
Encompass_Preferred
HY-100742A 10mg (R)-GNE-140 CAS: 2003234-63-5 (R)-GNE-140 is a potent lactate dehydrogenase A (LDHA) inhibitor, with IC50s of 3 nM and 5 nM for LDHA and LDHB, respectively; (R)-GNE-140 is 18-fold more potent than S enantiomer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9419