Learn More
TARGETMOL CHEMICALS INC R-Gossypol acetic acid 25MG
SDP
Catalog No. 502876885
Encompass_Preferred
Also available in 1 g, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. (R)-(-)-Gossypol acetic acid (AT101 acetate), the R-(-) enantiomer of Gossypol acetic acid, binds with Bcl-2, Bcl-xL, and Mcl-1 with Ki values of 0.32 uM, 0.48 uM, and 0.18 uM, respectively; it does not inhibit the BIR3 domain or BID. [Phase 2] Purity 96.73%
Catalog No. 50-287-6885 Supplier TARGETMOL CHEMICALS INC Supplier No. T639625MG
Also available in 1 g, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. (R)-(-)-Gossypol acetic acid (AT101 acetate), the R-(-) enantiomer of Gossypol acetic acid, binds with Bcl-2, Bcl-xL, and Mcl-1 with Ki values of 0.32 uM, 0.48 uM, and 0.18 uM, respectively; it does not inhibit the BIR3 domain or BID. [Phase 2] Purity 96.73%