Learn More
Medchemexpress LLC (R)-GSK-3685032 | 2170140-50-6 | >99.0% | 420.53 | 100 MG
SDP
Catalog No. 5000350060
Encompass_Preferred
(R)-GSK-3685032 is the R-enantiomer of GSK-3685032. It is a first-in-class, non-time-dependent, noncovalently, and reversible DNMT1-selective inhibitor, with an IC50 of 0.036 μM. It induces robust loss of DNA methylation, transcriptional activation, and inhibition of cancer cell growth.
  • Reversible DNMT1-selective inhibitor
  • Induces loss of DNA methylation
  • Promotes transcriptional activation
  • Inhibits cancer cell growth
Catalog No. 50-003-50060 Supplier Medchemexpress LLC Supplier No. HY139664A100MG
(R)-GSK-3685032 is the R-enantiomer of GSK-3685032. It is a first-in-class, non-time-dependent, noncovalently, and reversible DNMT1-selective inhibitor, with an IC50 of 0.036 μM. It induces robust loss of DNA methylation, transcriptional activation, and inhibition of cancer cell growth.
  • Reversible DNMT1-selective inhibitor
  • Induces loss of DNA methylation
  • Promotes transcriptional activation
  • Inhibits cancer cell growth