Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-GSK-3685032 | 2170140-50-6 | 99.0% | 420.53 | 50 MG
SDP

Catalog No. 5000349285
Encompass_Preferred

(R)-GSK-3685032 is the R-enantiomer of GSK-3685032, a first-in-class, non-time-dependent, noncovalently, reversible DNMT1-selective inhibitor with an IC50 of 0.036 μM. It is designed to induce robust loss of DNA methylation, transcriptional activation, and inhibition of cancer cell growth.

  • Selective DNMT1 inhibitor
  • Target: DNMT1
  • Induces loss of DNA methylation and transcriptional activation
  • Inhibits cancer cell growth
  • Solid storage: 4°C, sealed, away from moisture and light
  • Solvent storage: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture and light)

Catalog No. 50-003-49285 Supplier Medchemexpress LLC Supplier No. HY139664A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-GSK-3685032 is the R-enantiomer of GSK-3685032, a first-in-class, non-time-dependent, noncovalently, reversible DNMT1-selective inhibitor with an IC50 of 0.036 μM. It is designed to induce robust loss of DNA methylation, transcriptional activation, and inhibition of cancer cell growth.

  • Selective DNMT1 inhibitor
  • Target: DNMT1
  • Induces loss of DNA methylation and transcriptional activation
  • Inhibits cancer cell growth
  • Solid storage: 4°C, sealed, away from moisture and light
  • Solvent storage: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture and light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.