Learn More
Medchemexpress LLC (R)-GSK-3685032 | 2170140-50-6 | 99.0% | 420.53 g·mol⁻¹ | C22H24N6OS | 5 MG
SDP
Catalog No. 5000221304
Encompass_Preferred
(R)-GSK-3685032 is the R-enantiomer of GSK-3685032, a reversible, DNMT1-selective inhibitor used in epigenetics research to induce loss of DNA methylation, transcriptional activation, and cancer cell growth inhibition. The compound is characterized by a defined molecular formula and high reported purity, and it is supplied for in vitro and preclinical research use.
  • Reversible DNMT1-selective inhibitor with reported low-nanomolar activity.
  • Induces loss of DNA methylation and downstream transcriptional activation.
  • High reported purity and fully characterized molecular properties.
  • Available in small, research-scale pack sizes for preclinical studies.
Catalog No. 50-002-21304 Supplier Medchemexpress LLC Supplier No. HY139664A5MG
(R)-GSK-3685032 is the R-enantiomer of GSK-3685032, a reversible, DNMT1-selective inhibitor used in epigenetics research to induce loss of DNA methylation, transcriptional activation, and cancer cell growth inhibition. The compound is characterized by a defined molecular formula and high reported purity, and it is supplied for in vitro and preclinical research use.
  • Reversible DNMT1-selective inhibitor with reported low-nanomolar activity.
  • Induces loss of DNA methylation and downstream transcriptional activation.
  • High reported purity and fully characterized molecular properties.
  • Available in small, research-scale pack sizes for preclinical studies.