Learn More
Medchemexpress LLC (R)-GSK-3685032 | 2170140-50-6 | 99.0% | C22H24N6OS | 10MG
SDP
Catalog No. 5000258314
Encompass_Preferred
(R)-GSK-3685032 is the R-enantiomer of a reversible, DNMT1-selective small-molecule inhibitor supplied for research applications. It is provided as a purified solid intended for in vitro and exploratory studies and is not for human or clinical use.
  • Reversible DNMT1-selective inhibition suitable for epigenetic research.
  • High purity (98.97%).
  • Provided as a solid in a 10 mg package.
  • Storage recommendations for stock solutions included in datasheet.
  • Suitable for in vitro and mechanistic studies of DNA methylation.
Catalog No. 50-002-58314 Supplier Medchemexpress LLC Supplier No. HY139664A10MG
(R)-GSK-3685032 is the R-enantiomer of a reversible, DNMT1-selective small-molecule inhibitor supplied for research applications. It is provided as a purified solid intended for in vitro and exploratory studies and is not for human or clinical use.
  • Reversible DNMT1-selective inhibition suitable for epigenetic research.
  • High purity (98.97%).
  • Provided as a solid in a 10 mg package.
  • Storage recommendations for stock solutions included in datasheet.
  • Suitable for in vitro and mechanistic studies of DNA methylation.