Learn More
Bellco Glass, Inc. R/H Stainless Steel Pitch Blade Assembly , 10L For 10L Bioreator Systems
SDP
Catalog No. 501944733
Encompass_Preferred
Stainless steel pitch blade for 10l bioreactor systems . MADE IN THE USA Right hand upward. Material: 316L SST with Electropolish finish OD: 3/1/4"
Catalog No. 50-194-4733 Supplier Bellco Glass, Inc. Supplier No. 196402210
Stainless steel pitch blade for 10l bioreactor systems . MADE IN THE USA Right hand upward. Material: 316L SST with Electropolish finish OD: 3/1/4"