Learn More
R&D Systems™ Human Connexin 30.1/GJB5 (NP_005259) VersaClone cDNA
Catalog No. RDC1065
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
RDC1065 10 μg
1 options
This item is not returnable. View return policy
Catalog No. RDC1065 Supplier R&D Systems™ Supplier No. RDC1065
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
CX31.1; GJB5
The Human Connexin 30.1/GJB5 (NP_005259) VersaClone cDNA clone contains the complete ORF for Connexin 30.1/GJB5, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Connexin 30.1/GJB5 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
Specifications
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans