Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Fibronectin Quantikine ELISA Kit
SDP

Catalog No. DFBN10
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DFBN10 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DFBN10 Supplier R&D Systems™ Supplier No. DFBN10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

a 4.5 hour solid-phase ELISA designed to measure human SDF-1 alpha in cell culture supernates and platelet-poor plasma.

The Human Fibronectin Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Fibronectin in cell culture supernates (50 ul), cell lysates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), citrate plasma (10 ul), saliva (17 ul), urine (17 ul), human milk (10 ul).

Specifications

Assay Range 3.13 - 200ng/mL
Assay Sensitivity 0.579ng/mL
Detectable Analytes Fibronectin
Assay Solid Phase Sandwich ELISA
Conjugate HRP
Cross Reactivity < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well Strip Plate
Sample Type Cell Culture Supernatant, Cell Lysates, Human Milk, Plasma (EDTA, Citrate, Heparin), Saliva, Serum, Urine
Specificity Natural and Recombinant Human Fibronectin and Fibronectin fragments aa 1-1908, aa 607-1265, aa 627-702. Fibronectin fragment aa 631-705 cross-reacts approximately 0.415% Fibronectin fragments aa 1266-1908, aa 1722-1811, and aa 1913-2477 are not detected in this kit.
For Use With (Application) ELISA
Gene ID (Entrez) 2335
Host Species Human
Measuring Range 3.13-200ng/mL
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Cell Lysates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Citrate Plasma (10uL),Saliva (17uL),Urine (17uL),Human Milk (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.