Learn More
R&D Systems™ Human IL-18 R alpha/IL-1 R5 (NP_003846) VersaClone cDNA
Catalog No. RDC1117
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
RDC1117 10 μg
1 options
This item is not returnable. View return policy
Catalog No. RDC1117 Supplier R&D Systems™ Supplier No. RDC1117
Add to Cart
Add to Cart
This item is not returnable. View return policy
Interleukin 18 Receptor alpha
The Human IL-18 R alpha/IL-1 R5 (NP_003846) VersaClone cDNA clone contains the complete ORF for IL-18R alpha/IL-1 R5, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The IL-18R alpha/IL-1 R5 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
Specifications
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans