Learn More
Medchemexpress LLC HY-12693 10mg , R(+)-IAA-94 CAS:54197-31-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1269310MG
Encompass_Preferred
HY-12693 10mg R(+)-IAA-94 CAS: 54197-31-8 R(+)-IAA-94 (R(+)-Methylindazone) is a potent indanyloxyacetic acid blocker of epithelial chloride channels. R(+)-IAA-94 inhibits Nef-sdAb19 (single-domain antibody) interaction and binds to negative factor (Nef). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0656