Learn More
Medchemexpress LLC (R)-(-)-Ibuprofen-d3 | 121702-86-1 | 99.96% | C13H15D3O2 | 5 MG
SDP
Catalog No. 5000434852
Encompass_Preferred
(R)-(-)-Ibuprofen-d3 is a deuterium labeled form of (R)-(-)-Ibuprofen, an R enantiomer of Ibuprofen. Inactive on COX, it inhibits NF-κB activation and exhibits anti-inflammatory and antinociceptive effects. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules as tracers for quantitation during drug development, with potential to affect pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-34852 Supplier Medchemexpress LLC Supplier No. HY78131BS5MG
(R)-(-)-Ibuprofen-d3 is a deuterium labeled form of (R)-(-)-Ibuprofen, an R enantiomer of Ibuprofen. Inactive on COX, it inhibits NF-κB activation and exhibits anti-inflammatory and antinociceptive effects. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules as tracers for quantitation during drug development, with potential to affect pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS