Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-IL-17 modulator 4 | 2446804-29-9 | 99.6% | C27H34N6O2 | 1 ML
SDP

Catalog No. 5000353940
Encompass_Preferred

(R)-IL-17 modulator 4 is the R-configure of IL-17 modulator 4. It is a proagent of IL-17 modulator 1, which is an orally active, highly efficacious IL-17 modulator. This product is promising for research into IL-17A mediated diseases, including inflammation, autoimmune diseases, infectious diseases, cancer, and precancerous syndrome. It is for research use only and not sold to patients.

  • R-enantiomer of IL-17 modulator 4
  • Functions as a proagent of IL-17 modulator 1
  • Orally active and highly efficacious IL-17 modulator
  • Potentially useful for research into IL-17A mediated diseases
  • Supports studies on inflammation, autoimmune diseases, infectious diseases, cancer, and precancerous syndrome
  • For research use only
  • High purity of 99.6%
  • Molecular weight of 474.60
  • Chemical formula C27H34N6O2
  • Solid appearance
  • Off-white to gray color
  • Targets IL-17

Catalog No. 50-003-53940 Supplier Medchemexpress LLC Supplier No. HY141692B10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-IL-17 modulator 4 is the R-configure of IL-17 modulator 4. It is a proagent of IL-17 modulator 1, which is an orally active, highly efficacious IL-17 modulator. This product is promising for research into IL-17A mediated diseases, including inflammation, autoimmune diseases, infectious diseases, cancer, and precancerous syndrome. It is for research use only and not sold to patients.

  • R-enantiomer of IL-17 modulator 4
  • Functions as a proagent of IL-17 modulator 1
  • Orally active and highly efficacious IL-17 modulator
  • Potentially useful for research into IL-17A mediated diseases
  • Supports studies on inflammation, autoimmune diseases, infectious diseases, cancer, and precancerous syndrome
  • For research use only
  • High purity of 99.6%
  • Molecular weight of 474.60
  • Chemical formula C27H34N6O2
  • Solid appearance
  • Off-white to gray color
  • Targets IL-17

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.