Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-IL-17 modulator 4 | 2446804-29-9 | 99.6% | C27H34N6O2 | 10MG
SDP

Catalog No. 5000259693
Encompass_Preferred

(R)-IL-17 modulator 4 is a chiral small-molecule research compound used to modulate the IL-17 pathway in preclinical studies of inflammation, autoimmune conditions, infectious disease, and cancer. Supplied as an off-white to gray solid, it is characterized by high chemical purity and enantiomeric excess and is intended for laboratory research use.

  • High purity (99.56%) and enantiomeric excess (96.58%).
  • Molecular formula C27H34N6O2; molecular weight 474.60 Da.
  • Off-white to gray solid appearance suitable for handling in research labs.
  • Stable as a powder under recommended cold storage conditions.
  • Available in multiple small pack sizes to support experimental workflows.
  • Documentation available: data sheet, certificate of analysis, and SDS.

Catalog No. 50-002-59693 Supplier Medchemexpress LLC Supplier No. HY141692B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-IL-17 modulator 4 is a chiral small-molecule research compound used to modulate the IL-17 pathway in preclinical studies of inflammation, autoimmune conditions, infectious disease, and cancer. Supplied as an off-white to gray solid, it is characterized by high chemical purity and enantiomeric excess and is intended for laboratory research use.

  • High purity (99.56%) and enantiomeric excess (96.58%).
  • Molecular formula C27H34N6O2; molecular weight 474.60 Da.
  • Off-white to gray solid appearance suitable for handling in research labs.
  • Stable as a powder under recommended cold storage conditions.
  • Available in multiple small pack sizes to support experimental workflows.
  • Documentation available: data sheet, certificate of analysis, and SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.