Learn More
Medchemexpress LLC (R)-IL-17 modulator 4 | 2446804-29-9 | 99.6% | C27H34N6O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY141692B50MG
Encompass_Preferred
(R)-IL-17 modulator 4 is the R-configure of IL-17 modulator 4, a proagent of IL-17 modulator 1. IL-17 modulator 1 is an orally active and highly efficacious IL-17 modulator. (R)-IL-17 modulator 4 shows promise for research related to IL-17A mediated diseases such as inflammation, autoimmune diseases, infectious diseases, cancer, and precancerous syndrome.
  • R-configure of IL-17 modulator 4
  • Proagent of IL-17 modulator 1
  • Orally active and highly efficacious
  • Target: IL-17
  • Suitable for research on IL-17A mediated diseases, including inflammation, autoimmune diseases, infectious diseases, cancer, and precancerous syndrome
Catalog No. 50-003-56141