Learn More
Medchemexpress LLC HY-101354 5mg Medchemexpress, R-IMPP CAS:2133832-83-2 Purity:>98%
SDP
Catalog No. 501967833
Encompass_Preferred
Medchemexpress, HY-101354 5mg R-IMPP CAS:2133832-83-2 R-IMPP is an anti-secretagogue of PCSK9 (IC50=4.8 μM), which targets the 80S ribosome to inhibit PCSK9 protein translation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7833 Supplier Medchemexpress LLC Supplier No. HY1013545MG
Medchemexpress, HY-101354 5mg R-IMPP CAS:2133832-83-2 R-IMPP is an anti-secretagogue of PCSK9 (IC50=4.8 μM), which targets the 80S ribosome to inhibit PCSK9 protein translation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.