Learn More
Medchemexpress LLC (R)-INCB054329 | 1628607-63-5 | 98.9% | 348.36 g·mol⁻¹ | C19H16N4O3 | 5 MG
SDP
Catalog No. 5000262981
Encompass_Preferred
(R)-INCB054329 is the R-enantiomer of INCB054329, a potent BET bromodomain inhibitor used as an experimental control in biochemical and cellular research. Supplied as a powder for research use, it has molecular formula C19H16N4O3 and molecular weight 348.36 g·mol⁻1. Typical package sizes range from 1 mg to 100 mg, and analytical documentation (COA/LCMS) is available from the supplier.
Catalog No. 50-002-62981 Supplier Medchemexpress LLC Supplier No. HY112504D5MG
(R)-INCB054329 is the R-enantiomer of INCB054329, a potent BET bromodomain inhibitor used as an experimental control in biochemical and cellular research. Supplied as a powder for research use, it has molecular formula C19H16N4O3 and molecular weight 348.36 g·mol⁻1. Typical package sizes range from 1 mg to 100 mg, and analytical documentation (COA/LCMS) is available from the supplier.