Learn More
Strem, An Ascensus Company CAS 164858-78-0. 50mg. (R)-()-2-[2-(Diphenylphosphino)phenyl]-4-(1-methylethyl)-45-dihydrooxazole 98% (R)-iPr-PHOX. MFCD02684553
SDP
Catalog No. 502110757
Encompass
CAS 164858-78-0. 50mg. (R)-()-2-[2-(Diphenylphosphino)phenyl]-4-(1-methylethyl)-45-dihydrooxazole 98% (R)-iPr-PHOX. MFCD02684553. Molecular Weight 373.44. Molecular Formula C24H24NOP. Color/form white pwdr. Strem 15-1821. http//www.strem.com/catalog/v/15-1821/
Catalog No. 50-211-0757 Supplier Strem, An Ascensus Company Supplier No. 15182150MG
CAS 164858-78-0. 50mg. (R)-()-2-[2-(Diphenylphosphino)phenyl]-4-(1-methylethyl)-45-dihydrooxazole 98% (R)-iPr-PHOX. MFCD02684553. Molecular Weight 373.44. Molecular Formula C24H24NOP. Color/form white pwdr. Strem 15-1821. http//www.strem.com/catalog/v/15-1821/