Learn More
Selleck Chemical LLC (R)-(-)-JQ1 Enantiomer S2965-5mg
SDP
Catalog No. 503111692
Encompass_Preferred
(R)-(-)-JQ1 Enantiomer is the stereoisomer of ( )-JQ1 ( )-JQ1 potently decreases expression of both BRD4 target genes whereas (R)-(-)-JQ1 Enantiomer has no effect
Catalog No. 50-311-1692 Supplier Selleck Chemical LLC Supplier No. S29655MG
(R)-(-)-JQ1 Enantiomer is the stereoisomer of ( )-JQ1 ( )-JQ1 potently decreases expression of both BRD4 target genes whereas (R)-(-)-JQ1 Enantiomer has no effect