Learn More
Selleck Chemical LLC (R)-(-)-JQ1 Enantiomer S2965-25mg
SDP
Catalog No. 503112428
Encompass_Preferred
(R)-(-)-JQ1 Enantiomer is the stereoisomer of ( )-JQ1 ( )-JQ1 potently decreases expression of both BRD4 target genes whereas (R)-(-)-JQ1 Enantiomer has no effect
Catalog No. 50-311-2428 Supplier Selleck Chemical LLC Supplier No. S296525MG
(R)-(-)-JQ1 Enantiomer is the stereoisomer of ( )-JQ1 ( )-JQ1 potently decreases expression of both BRD4 target genes whereas (R)-(-)-JQ1 Enantiomer has no effect