Learn More
Medchemexpress LLC HY-B0580B 5mg Medchemexpress, (R)-Ketorolac CAS:66635-93-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0580B5MG
Encompass_Preferred
Medchemexpress, HY-B0580B 5mg (R)-Ketorolac CAS:66635-93-6 (R)-Ketorolac is the R-enantiomer of Ketorolac, shows potent analgesic activity, reduces ulcerogenic potential. (R)-Ketorolac is inactive on COX [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2922