Learn More
Medchemexpress LLC (R)-Ketorolac | 66635-93-6 | 99.65% | 50 MG
SDP
Catalog No. 5000409028
Encompass_Preferred
(R)-Ketorolac is an orally active inhibitor of Cdc42 and Rac1 GTPases. It has been shown to alter ovarian cancer cell behaviors crucial for invasion and metastasis and can ameliorate cancer-associated cachexia.
  • Orally active Cdc42 and Rac1 inhibitor.
  • Inhibits GTPase.
  • Alters ovarian cancer cell behaviors critical for invasion and metastasis.
  • Ameliorates cancer-associated cachexia.
Catalog No. 50-004-09028 Supplier Medchemexpress LLC Supplier No. HYB0580B50MG
(R)-Ketorolac is an orally active inhibitor of Cdc42 and Rac1 GTPases. It has been shown to alter ovarian cancer cell behaviors crucial for invasion and metastasis and can ameliorate cancer-associated cachexia.
  • Orally active Cdc42 and Rac1 inhibitor.
  • Inhibits GTPase.
  • Alters ovarian cancer cell behaviors critical for invasion and metastasis.
  • Ameliorates cancer-associated cachexia.