Learn More
Medchemexpress LLC (R)-Ketorolac | 66635-93-6 | 99.7% | 10 MM 1 ML
SDP
Catalog No. 5000408461
Encompass_Preferred
(R)-Ketorolac is an orally active inhibitor of Cdc42 and Rac1. It inhibits GTPase and modifies ovarian cancer cell behaviors crucial for invasion and metastasis. This compound also ameliorates cancer-associated cachexia.
  • Inhibits Cdc42 and Rac1
  • Inhibits GTPase activity
  • Modifies ovarian cancer cell behaviors for invasion and metastasis
  • Ameliorates cancer-associated cachexia
  • Decreases filopodia number and length
  • Reduces cell adherence to fibronectin and collagen
  • Prolongs survival in C26 tumour-bearing mice
  • Alleviates C26-induced weight loss
  • Prevents C26-induced T-lymphopenia
Catalog No. 50-004-08461 Supplier Medchemexpress LLC Supplier No. HYB058010MM/1ML
(R)-Ketorolac is an orally active inhibitor of Cdc42 and Rac1. It inhibits GTPase and modifies ovarian cancer cell behaviors crucial for invasion and metastasis. This compound also ameliorates cancer-associated cachexia.
  • Inhibits Cdc42 and Rac1
  • Inhibits GTPase activity
  • Modifies ovarian cancer cell behaviors for invasion and metastasis
  • Ameliorates cancer-associated cachexia
  • Decreases filopodia number and length
  • Reduces cell adherence to fibronectin and collagen
  • Prolongs survival in C26 tumour-bearing mice
  • Alleviates C26-induced weight loss
  • Prevents C26-induced T-lymphopenia